Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008966.1 | NP_001008966.1 | 1 | 98 | ||
GenBank mRNA | AY510353.1 | - | 1 | 98 |
Gene Symbol: LOC103691019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758872.1 | XP_008757094.1 | 1 | 98 |
Gene Symbol: Vom1r63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008967.1 | NP_001008967.1 | 1 | 98 | ||
GenBank mRNA | AY510345.1 | - | 1 | 98 |