Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 12 | 1910 | ||
GenBank mRNA | AK299293.1 | - | 12 | 1869 | ||
AK301461.1 | - | 11 | 1678 | |||
AK310703.1 | - | 12 | 1851 | |||
AK310938.1 | - | 10 | 1410 | |||
AL833263.1 | - | 11 | 1633 | |||
BC033903.1 | - | 12 | 1892 | |||
BC107880.1 | - | 12 | 1891 |