Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009535.1 | NP_001009535.1 | 9 | 1743 | ||
GenBank mRNA | BC061722.1 | - | 9 | 1743 | 5598508 |