Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 3 | 542 | ||
NM_001159739.2 | NP_001153211.1 | 4 | 512 | |||
NM_001287025.1 | NP_001273954.1 | 3 | 404 | |||
GenBank mRNA | BC100080.1 | - | 4 | 369 | 7324340 | |
CO566642.1 | - | 4 | 375 | |||
DY308874.1 | - | 4 | 436 | |||
DY573045.1 | - | 2 | 237 | |||
FQ210706.1 | - | 3 | 371 | |||
FQ218526.1 | - | 4 | 385 | |||
FQ218726.1 | - | 4 | 379 | |||
FQ219557.1 | - | 4 | 379 | |||
FQ219615.1 | - | 4 | 512 | |||
FQ219704.1 | - | 4 | 379 | |||
FQ231428.1 | - | 4 | 383 | |||
S72506.1 | - | 3 | 390 | |||
S82820.1 | - | 3 | 542 | |||
X78847.1 | - | 3 | 390 |