Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001396.2 | NP_001001396.1 | 17 | 3633 | ||
NM_001684.4 | NP_001675.3 | 17 | 3633 | |||
GenBank mRNA | AK026443.1 | - | 5 | 659 | ||
AK292458.1 | - | 16 | 2972 | |||
BC140774.1 | - | 17 | 3305 | 9021642 | ||
BC150293.1 | - | 16 | 2990 | 8860318 | ||
BX537444.1 | - | 17 | 3860 | |||
CR749393.1 | - | 17 | 3563 | |||
CR936708.1 | - | 17 | 3588 | |||
M25874.1 | - | 16 | 3133 | |||
M83363.1 | - | 16 | 3133 |