Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031502.1 | NP_113690.1 | 1144 | |||
GenBank mRNA | FQ233367.1 | - | 8 | 1139 | ||
J00703.2 | - | 1144 |
Gene Symbol: LOC103689940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761448.1 | XP_008759670.1 | 8 | 1153 |