Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135669.1 | NP_001129141.1 | 1, 2 | 263 | ||
NM_004736.3 | NP_004727.2 | 1, 2 | 263 | |||
GenBank mRNA | AF089744.1 | - | 1, 2 | 236 | ||
AF099082.1 | - | 1, 2 | 240 | |||
AF115389.1 | - | 1, 2 | 71 | |||
AK309750.1 | - | 1, 2 | 251 | |||
BC041142.1 | - | 1, 2 | 241 | 5582839 | ||
DB461474.1 | - | 1, 2 | 263 |
Set Membership: |
G-protein coupled receptor |