Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017473.1 | NP_001017473.1 | 2 | 256 | ||
XM_006246205.1 | XP_006246267.1 | 2 | 284 | |||
GenBank mRNA | BC093386.1 | - | 2 | 256 | 7389386 |
Gene Symbol: LOC103689927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246315.1 | XP_006246377.1 | 2 | 284 | ||
XM_008767740.1 | XP_008765962.1 | 2 | 314 |