Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025134.1 | NP_001020305.1 | 2 | 321 | ||
XM_006246316.2 | XP_006246378.1 | 3 | 301 | |||
GenBank mRNA | AY323237.1 | - | 2 | 321 | ||
FQ209456.1 | - | 3 | 242 | |||
FQ209915.1 | - | 3 | 241 | |||
FQ210236.1 | - | 3 | 241 | |||
FQ210461.1 | - | 3 | 241 | |||
FQ210609.1 | - | 3 | 241 | |||
FQ210647.1 | - | 3 | 241 | |||
FQ210681.1 | - | 3 | 240 | |||
FQ211104.1 | - | 3 | 240 | |||
FQ211208.1 | - | 3 | 241 | |||
FQ211225.1 | - | 3 | 239 | |||
FQ211785.1 | - | 3 | 241 | |||
FQ216941.1 | - | 3 | 249 | |||
FQ216945.1 | - | 3 | 240 | |||
FQ216963.1 | - | 3 | 243 | |||
FQ217023.1 | - | 3 | 239 | |||
FQ217097.1 | - | 3 | 241 | |||
FQ217180.1 | - | 3 | 241 | |||
FQ217379.1 | - | 3 | 240 | |||
FQ217445.1 | - | 3 | 250 | |||
FQ217512.1 | - | 3 | 242 | |||
FQ217598.1 | - | 3 | 241 | |||
FQ217789.1 | - | 3 | 242 | |||
FQ217857.1 | - | 3 | 243 | |||
FQ217947.1 | - | 3 | 239 | |||
FQ217992.1 | - | 3 | 241 | |||
FQ218025.1 | - | 3 | 242 | |||
FQ220995.1 | - | 3 | 241 | |||
FQ222364.1 | - | 3 | 240 | |||
FQ222807.1 | - | 3 | 240 | |||
FQ223593.1 | - | 3 | 242 | |||
FQ223655.1 | - | 3 | 240 | |||
FQ223864.1 | - | 3 | 241 | |||
FQ223921.1 | - | 3 | 241 | |||
FQ223972.1 | - | 3 | 242 | |||
FQ223991.1 | - | 3 | 243 | |||
FQ224012.1 | - | 3 | 241 | |||
FQ224065.1 | - | 3 | 242 | |||
FQ224075.1 | - | 3 | 240 | |||
FQ224142.1 | - | 3 | 241 | |||
FQ224210.1 | - | 3 | 242 | |||
FQ224267.1 | - | 3 | 242 | |||
FQ224372.1 | - | 3 | 251 | |||
FQ224703.1 | - | 3 | 243 | |||
FQ229024.1 | - | 3 | 242 |