Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC497940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017476.1 | NP_001017476.1 | 4 | 798 | ||
XM_006246777.2 | XP_006246839.1 | 4 | 945 | |||
XM_006246778.2 | XP_006246840.1 | 4 | 949 | |||
XM_006246781.2 | XP_006246843.1 | 4 | 946 | |||
XM_006246782.2 | XP_006246844.1 | 4 | 942 | |||
GenBank mRNA | BC079326.1 | - | 4 | 798 | 7135484 |