Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246868.2 | XP_006246930.1 | 4 | 1355 | ||
XM_006246869.2 | XP_006246931.1 | 5 | 1253 | |||
XM_006246870.2 | XP_006246932.1 | 5 | 1160 | |||
XM_573142.6 | XP_573142.4 | 4 | 1355 | |||
GenBank mRNA | FQ219135.1 | - | 4 | 827 |