Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam222b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109038.1 | NP_001102508.1 | 3 | 315 | ||
XM_006246944.1 | XP_006247006.1 | 3 | 334 | |||
XM_008768094.1 | XP_008766316.1 | 2 | 812 | |||
XM_008768095.1 | XP_008766317.1 | 3 | 945 |