Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191924.1 | NP_001178853.1 | 11 | 3198 | ||
GenBank mRNA | FQ225006.1 | - | 10 | 1844 |