Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173471.1 | NP_001166942.1 | 3 | 234 | ||
XM_008768097.1 | XP_008766319.1 | 3 | 257 | |||
GenBank mRNA | BC167116.1 | - | 3 | 234 | 6922274 |