Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004739.3 | NP_004730.2 | 13 | 1582 | ||
XM_017018561.1 | XP_016874050.1 | 13 | 1434 | |||
GenBank mRNA | AB016591.1 | - | 13 | 1515 | ||
AF295807.1 | - | 13 | 1192 | |||
AK301569.1 | - | 13 | 1417 | |||
BC023656.2 | - | 13 | 1378 | |||
BC053650.1 | - | 13 | 1400 | 5531965 | ||
CR749481.1 | - | 11 | 1103 |