Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100787.1 | NP_001094257.1 | 2 | 873 | ||
GenBank mRNA | BC161863.1 | - | 2 | 873 | 7098209 |