Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284388.1 | NP_001271317.1 | 3 | 429 | ||
NM_001284389.1 | NP_001271318.1 | 3 | 432 | |||
NM_004741.4 | NP_004732.2 | 3 | 429 | |||
XM_005270273.1 | XP_005270330.1 | 3 | 406 | |||
GenBank mRNA | AK225101.1 | - | 3 | 219 | ||
AK314363.1 | - | 3 | 216 | |||
AY820769.1 | - | 3 | 197 | |||
BC001883.1 | - | 3 | 204 | 3533568 | ||
D21262.1 | - | 3 | 190 | |||
DA588285.1 | - | 3 | 429 | |||
Z34289.1 | - | 3 | 248 |