Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLGAP1 SIRNA Assays ›
Gene Symbol:
DLGAP1
Gene Name:
DLG associated protein 1
Gene Aliases:
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
Chromosome Location:
Chr.18: 3496032 - 4455307 on Build GRCh38
UniGene ID:
Hs.594640
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17645
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLGAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003809.2
NP_001003809.1
7
1493
NM_001242761.1
NP_001229690.1
10
2850
NM_001242762.1
NP_001229691.1
7
1436
NM_001242763.1
NP_001229692.1
8
1550
NM_001242764.1
NP_001229693.1
7
1466
NM_001242765.1
NP_001229694.1
7
1493
NM_001242766.1
NP_001229695.1
8
1523
NM_001308390.1
NP_001295319.1
8
1548
NM_004746.3
NP_004737.2
10
2850
XM_005258171.2
XP_005258228.1
11
2921
XM_005258172.1
XP_005258229.1
11
2916
XM_005258173.3
XP_005258230.2
9
1712
XM_005258174.4
XP_005258231.1
8
1769
XM_006722367.3
XP_006722430.1
9
1880
XM_017026080.1
XP_016881569.1
8
1849
XM_017026081.1
XP_016881570.1
7
1513
XM_017026082.1
XP_016881571.1
8
1668
XM_017026083.1
XP_016881572.1
7
1738
XM_017026084.1
XP_016881573.1
7
1735
XM_017026085.1
XP_016881574.1
7
1728
GenBank mRNA
AB000276.1
-
7
1436
AB000277.1
-
7
2390
AK294717.1
-
7
1436
AK294747.1
-
8
1523
AK294777.1
-
8
1473
AK299880.1
-
7
1466
AK304101.1
-
10
2832
AK316099.1
-
10
2850
BC136453.1
-
7
2382
9020442
BC136454.1
-
7
2382
9020443
U67988.1
-
8
1440
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein binding
protein domain specific binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0