Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109057.1 | NP_001102527.1 | 7 | 924 | ||
XM_008768826.1 | XP_008767048.1 | 1 | 594 |