Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139114.3 | NP_620814.1 | 4 | 516 | ||
GenBank mRNA | BC078724.1 | - | 3 | 455 | 7125608 | |
FQ210520.1 | - | 4 | 471 | |||
FQ217417.1 | - | 4 | 472 | |||
FQ217514.1 | - | 4 | 475 | |||
FQ218767.1 | - | 4 | 471 | |||
FQ221301.1 | - | 4 | 484 | |||
FQ221436.1 | - | 4 | 473 | |||
FQ221541.1 | - | 4 | 472 | |||
FQ221627.1 | - | 4 | 475 | |||
FQ221775.1 | - | 4 | 472 | |||
FQ221788.1 | - | 4 | 471 | |||
FQ221816.1 | - | 4 | 472 | |||
FQ222173.1 | - | 4 | 472 | |||
FQ222227.1 | - | 4 | 475 | |||
FQ222322.1 | - | 4 | 471 | |||
FQ222450.1 | - | 4 | 485 | |||
FQ222725.1 | - | 4 | 472 | |||
FQ224390.1 | - | 4 | 471 | |||
FQ227511.1 | - | 4 | 486 | |||
FQ228660.1 | - | 3 | 460 | |||
FQ230911.1 | - | 4 | 475 | |||
X78167.1 | - | 3 | 453 |
Gene Symbol: RGD1565131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248902.2 | XP_006248964.1 | 2 | 467 | ||
XM_008769031.1 | XP_008767253.1 | 2 | 467 |