Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 8 | 495 | ||
NM_001012632.1 | NP_001012650.1 | 7 | 294 | |||
NM_001012633.1 | NP_001012651.1 | 7 | 428 | |||
NM_001012634.1 | NP_001012652.1 | 6 | 235 | |||
NM_001012635.1 | NP_001012653.1 | 6 | 230 | |||
NM_001012636.1 | NP_001012654.1 | 5 | 218 | |||
NM_001012718.1 | NP_001012736.1 | 7 | 431 | |||
NM_001308078.1 | NP_001295007.1 | 6 | 429 | |||
NM_004221.4 | NP_004212.4 | 7 | 428 | |||
GenBank mRNA | AK296143.1 | - | 6 | 240 | ||
AY495331.1 | - | 7 | 418 | |||
AY495332.1 | - | 7 | 418 | |||
AY495333.1 | - | 6 | 376 | |||
BC009401.2 | - | 6 | 566 | 4300407 | ||
BC018782.1 | - | 7 | 275 | 4873608 | ||
BC105602.1 | - | 7 | 296 | 6496101 | ||
BF568456.1 | - | 6 | 567 | |||
BG748458.1 | - | 7 | 374 | |||
BM759359.1 | - | 4 | 169 | |||
BM822755.1 | - | 8 | 403 | |||
EU339107.1 | - | 4 | 97 | |||
FJ985780.1 | - | 6 | 322 | |||
JN546102.1 | - | 5 | 157 | |||
M59807.1 | - | 6 | 414 |