Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131001.1 | NP_001124473.1 | 2 | 181 | ||
GenBank mRNA | BC160864.1 | - | 2 | 181 | 7324585 |