Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005098.3 | NP_005089.2 | 2 | 1657 | ||
GenBank mRNA | AF060154.1 | - | 2 | 1548 | ||
AF087036.1 | - | 2 | 1325 | |||
BC006313.2 | - | 2 | 1623 | 4123825 | ||
BC067827.1 | - | 2 | 1632 | 30336424 |