Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004750.4 | NP_004741.1 | 3 | 608 | ||
XM_011528422.2 | XP_011526724.1 | 3 | 690 | |||
XM_011528423.2 | XP_011526725.1 | 3 | 414 | |||
XM_011528424.2 | XP_011526726.1 | 3 | 682 | |||
GenBank mRNA | AF059293.1 | - | 3 | 532 | ||
AF073515.1 | - | 3 | 617 | |||
AF178684.1 | - | 3 | 465 | |||
AY358291.1 | - | 3 | 544 | |||
BC044634.1 | - | 3 | 541 | 5926400 |