Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004751.2 | NP_004742.1 | 3 | 1281 | ||
GenBank mRNA | AF038650.1 | - | 1 | 832 | ||
AF102542.1 | - | 2 | 1185 | |||
AK312852.1 | - | 2 | 1064 | |||
BC017032.1 | - | 3 | 1140 | 3851937 | ||
EF152283.1 | - | 3 | 1422 |