Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017497.1 | NP_001017497.1 | 3 | 440 | ||
GenBank mRNA | X77816.1 | - | 3 | 440 |