Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025138.1 | NP_001020309.1 | 2 | 182 | ||
XM_008770160.1 | XP_008768382.1 | 3 | 397 | |||
GenBank mRNA | BC091217.1 | - | 2 | 182 | 7384360 |