Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251412.1 | XP_006251474.1 | 4 | 1329 | ||
XM_006251414.2 | XP_006251476.1 | 4 | 1329 | |||
XM_008770382.1 | XP_008768604.1 | 4 | 1329 | |||
XM_008770383.1 | XP_008768605.1 | 4 | 888 | |||
XM_008770384.1 | XP_008768606.1 | 4 | 1329 |