Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025140.2 | NP_001020311.2 | 16 | 2213 | ||
XM_008770467.1 | XP_008768689.1 | 17 | 2529 | |||
GenBank mRNA | FM091221.1 | - | 2 | 155 |