Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128311.1 | NP_001121783.1 | 10 | 1925 |
Gene Symbol: Abcg3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014133.1 | NP_001014155.1 | 1871 | |||
GenBank mRNA | BC085773.1 | - | 1871 | 7096429 |
Gene Symbol: Abcg3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037205.1 | NP_001032282.1 | 16 | 1979 | ||
GenBank mRNA | BC100073.1 | - | 16 | 1979 | 7372627 |