Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109097.1 | NP_001102567.1 | 6 | 842 | ||
XM_006251655.2 | XP_006251717.1 | 6 | 889 | |||
GenBank mRNA | FQ214748.1 | - | 6 | 842 | ||
FQ214808.1 | - | 6 | 842 | |||
FQ215077.1 | - | 844 | ||||
FQ215190.1 | - | 6 | 844 | |||
FQ215360.1 | - | 6 | 848 | |||
FQ215382.1 | - | 5 | 678 | |||
FQ215901.1 | - | 6 | 846 | |||
FQ215918.1 | - | 6 | 843 | |||
FQ216019.1 | - | 6 | 882 | |||
FQ216101.1 | - | 6 | 844 | |||
FQ216248.1 | - | 6 | 843 | |||
FQ216396.1 | - | 6 | 826 | |||
FQ216753.1 | - | 6 | 826 | |||
FQ216842.1 | - | 6 | 826 | |||
FQ217380.1 | - | 864 | ||||
FQ224894.1 | - | 6 | 846 | |||
FQ224977.1 | - | 6 | 842 |