Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109105.1 | NP_001102575.1 | 3 | 406 | ||
XM_008770891.1 | XP_008769113.1 | 4 | 320 | |||
XM_008770892.1 | XP_008769114.1 | 2 | 313 | |||
GenBank mRNA | FQ223060.1 | - | 3 | 206 |