Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004225.2 | NP_004216.2 | 1 | 3211 | ||
XM_011543852.2 | XP_011542154.1 | 1 | 3185 | |||
GenBank mRNA | AB016816.1 | - | 1 | 2648 | ||
AK027007.1 | - | 1 | 1244 | |||
BC014226.2 | - | 1 | 1502 |