Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252757.2 | XP_006252819.1 | 5 | 2585 | ||
XM_008771158.1 | XP_008769380.1 | 6 | 3593 | |||
XM_573855.5 | XP_573855.2 | 6 | 2618 |