Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109107.2 | NP_001102577.2 | 5 | 921 | ||
GenBank mRNA | CB316326.1 | - | 3 | 427 | ||
CB780985.1 | - | 2 | 232 |