Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 750 | ||
NM_001270562.1 | NP_001257491.1 | 4 | 648 | |||
NM_001270563.1 | NP_001257492.1 | 4 | 608 | |||
NM_022593.4 | NP_072115.1 | 4 | 644 | |||
XM_006237724.2 | XP_006237786.1 | 4 | 645 | |||
GenBank mRNA | BC060566.1 | - | 4 | 619 | 6918038 | |
CK355540.1 | - | 4 | 644 | |||
FM093546.1 | - | 4 | 610 | |||
FM110992.1 | - | 4 | 614 | |||
FQ213660.1 | - | 4 | 658 | |||
FQ214805.1 | - | 4 | 750 |