Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004227.3 | NP_004218.1 | 8, 9 | 823 | ||
GenBank mRNA | AJ005197.1 | - | 8, 9 | 794 | ||
AJ223957.1 | - | 8, 9 | 744 | |||
AK295163.1 | - | 3, 4 | 615 | |||
AK313728.1 | - | 9, 10 | 839 | |||
BC008191.1 | - | 1, 2 | 62 | |||
BC028717.1 | - | 8, 9 | 859 | 4811560 |