Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 3, 4 | 679 | ||
NM_001292019.1 | NP_001278948.1 | 3, 4 | 245 | |||
NM_004762.3 | NP_004753.1 | 3, 4 | 282 | |||
NM_017456.3 | NP_059430.2 | 3, 4 | 282 | |||
XM_011525475.2 | XP_011523777.1 | 2, 3 | 141 | |||
XM_011525476.2 | XP_011523778.1 | 2, 3 | 141 | |||
XM_011525477.1 | XP_011523779.1 | 1, 2 | 60 | |||
XM_017025336.1 | XP_016880825.1 | 3, 4 | 352 | |||
XM_017025337.1 | XP_016880826.1 | 3, 4 | 237 | |||
XM_017025338.1 | XP_016880827.1 | 3, 4 | 668 | |||
GenBank mRNA | AK293894.1 | - | 3, 4 | 679 | ||
AK316075.1 | - | 3, 4 | 238 | |||
AK316277.1 | - | 3, 4 | 226 | |||
BC038385.1 | - | 3, 4 | 203 | 4755203 | ||
BC050452.1 | - | 3, 4 | 192 | 6043039 | ||
BX339155.2 | - | 3, 4 | 262 | |||
DA125333.1 | - | 3, 4 | 245 | |||
HY049953.1 | - | 3, 4 | 282 | |||
M85169.1 | - | 3, 4 | 232 |