Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 7 | 882 | ||
GenBank mRNA | BC061781.1 | - | 7 | 857 | ||
BC126087.1 | - | 7 | 882 | 8368624 | ||
FQ214607.1 | - | 7 | 968 | |||
FQ214670.1 | - | 7 | 891 | |||
FQ214880.1 | - | 7 | 895 | |||
FQ215213.1 | - | 6 | 797 | |||
FQ215606.1 | - | 6 | 770 | |||
FQ215810.1 | - | 7 | 895 | |||
FQ216226.1 | - | 6 | 820 | |||
FQ216307.1 | - | 7 | 781 | |||
FQ216584.1 | - | 7 | 894 | |||
FQ223237.1 | - | 7 | 896 | |||
FQ224880.1 | - | 7 | 893 | |||
FQ224920.1 | - | 7 | 897 | |||
FQ224970.1 | - | 7 | 890 | |||
L36250.1 | - | 7 | 881 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 7 | 1038 |