Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017293.1 | NP_058989.1 | 5 | 1074 | ||
GenBank mRNA | U70372.2 | - | 5 | 1074 | ||
X98374.1 | - | 5 | 951 |