Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170421.1 | NP_001163892.1 | 5 | 483 | ||
XM_008771753.1 | XP_008769975.1 | 5 | 691 | |||
XM_008771754.1 | XP_008769976.1 | 6 | 515 | |||
XM_008771755.1 | XP_008769977.1 | 5 | 487 | |||
XM_008771756.1 | XP_008769978.1 | 5 | 520 | |||
XM_008771757.1 | XP_008769979.1 | 5 | 874 | |||
XM_008771758.1 | XP_008769980.1 | 5 | 640 | |||
XM_008771759.1 | XP_008769981.1 | 5 | 429 | |||
XM_008771760.1 | XP_008769982.1 | 3 | 702 | |||
XM_008771761.1 | XP_008769983.1 | 4 | 322 | |||
GenBank mRNA | BC158594.1 | - | 5 | 483 | 7111099 | |
FQ225186.1 | - | 1 | 165 | |||
FQ231877.1 | - | 1 | 165 |
Gene Symbol: LOC103690053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771792.1 | XP_008770014.1 | 3 | 692 |