Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197244.1 | NP_001184173.1 | 3 | 337 | ||
NM_001301061.1 | NP_001287990.1 | 4 | 459 | |||
NM_001707.3 | NP_001698.2 | 3 | 337 | |||
NR_036682.1 | 3 | 320 | ||||
GenBank mRNA | AJ223979.1 | - | 3 | 216 | ||
AK290091.1 | - | 3 | 245 | |||
AL525884.3 | - | 4 | 400 | |||
AL553260.3 | - | 3 | 604 | |||
BC000956.2 | - | 3 | 296 | 3450946 | ||
BC001967.2 | - | 3 | 203 | 3461829 | ||
BC009548.1 | - | 3 | 247 | 3897805 | ||
BI837138.1 | - | 3 | 308 | |||
BQ109412.1 | - | 3 | 269 | |||
DA688517.1 | - | 3 | 320 | |||
X89985.1 | - | 3 | 217 |