Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109122.1 | NP_001102592.1 | 6 | 1087 | ||
XM_006254601.2 | XP_006254663.1 | 5 | 1054 | |||
GenBank mRNA | JN587497.1 | - | 4 | 731 |