Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574154.6 | XP_574154.2 | 1 | 751 |
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022539.1 | NP_071984.1 | 5 | 721 | ||
GenBank mRNA | BC070892.1 | - | 5 | 636 | 7099147 | |
BC091150.1 | - | 5 | 735 | 7122560 | ||
L10652.1 | - | 5 | 721 |