Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164267.1 | NP_001157739.1 | 9 | 1159 | ||
NM_005452.5 | NP_005443.3 | 9 | 1321 | |||
XM_017011484.1 | XP_016866973.1 | 9 | 1035 | |||
XM_017011485.1 | XP_016866974.1 | 7 | 705 | |||
XM_017011486.1 | XP_016866975.1 | 6 | 538 | |||
XM_017011487.1 | XP_016866976.1 | 7 | 705 | |||
GenBank mRNA | AF447870.1 | - | 9 | 1013 | ||
AK292171.1 | - | 9 | 1304 | |||
AK292616.1 | - | 9 | 1104 | |||
AK297925.1 | - | 9 | 916 | |||
AK298151.1 | - | 9 | 870 | |||
AK308534.1 | - | 8 | 819 | |||
AK311388.1 | - | 8 | 1018 | |||
AK315616.1 | - | 9 | 1033 | |||
BC000388.2 | - | 9 | 1008 | 2821198 | ||
BX439421.2 | - | 9 | 1008 |