Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC108823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012353.1 | NP_001012353.1 | 2 | 1184 | ||
GenBank mRNA | BC089836.1 | - | 2 | 1184 | 7326302 |
Gene Symbol: LOC100910934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751792.3 | XP_003751840.2 | 1 | 1288 |
Gene Symbol: LOC100910979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254758.2 | XP_006254820.1 | 2 | 1215 |
Gene Symbol: LOC102555392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254757.2 | XP_006254819.1 | 1066 |
Gene Symbol: LOC103694226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772196.1 | XP_008770418.1 | 1 | 1066 |
Gene Symbol: RGD1309362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024884.1 | NP_001020055.1 | 2 | 1288 | ||
XM_006254759.2 | XP_006254821.1 | 2 | 1590 | |||
GenBank mRNA | BC098065.1 | - | 2 | 1288 | 7458735 |