Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234386.2 | XP_006234448.1 | 6 | 1181 | ||
XM_006234388.2 | XP_006234450.1 | 6 | 421 | |||
XM_006234389.2 | XP_006234451.1 | 6 | 1124 | |||
XM_008761932.1 | XP_008760154.1 | 3 | 1181 |