Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024906.1 | NP_001020077.1 | 3 | 292 | ||
XM_006255120.2 | XP_006255182.1 | 3 | 372 | |||
XM_006255121.2 | XP_006255183.1 | 3 | 400 | |||
GenBank mRNA | BC097448.1 | - | 3 | 292 | 7456585 |