Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004767.3 | NP_004758.3 | 1 | 189 | ||
GenBank mRNA | AK222639.1 | - | 1 | 171 | ||
AK313044.1 | - | 1 | 216 | |||
BC050334.1 | - | 1 | 189 | 5174449 | ||
Y16280.1 | - | 1 | 177 |
Set Membership: |
G-protein coupled receptor |