Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004767.3 | NP_004758.3 | 2 | 709 | ||
XM_011510158.2 | XP_011508460.1 | 2 | 225 | |||
GenBank mRNA | AK222639.1 | - | 2 | 691 | ||
AK295576.1 | - | 4 | 692 | |||
AK313044.1 | - | 2 | 736 | |||
BC050334.1 | - | 2 | 709 | 5174449 | ||
Y16280.1 | - | 2 | 697 |
Set Membership: |
G-protein coupled receptor |